Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Manufactured from borosilicate glass
Brand:  Pyrex 1541/22D
 View more versions of this product
Product Code. 11377884
  • £485.00 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Compliance
ISO 4796
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20,000 mL
Borosilicate Glass
Aspirator Bottle
59 mm
492 mm
288 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products