missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Aspirator Bottle with Tubing Side Arm
Store and discharge a variety of liquids with this robust aspirator bottle. Pyrex™ Aspirator Bottles with Tubing Side Arm are useful as a delivery or storage container for a wide range of solutions. These bottles feature sturdy ring necks with a heavy duty rims.
Brand: Pyrex 1531/12D
319.20 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Tubular side arm for attachment of 9mm bore flexible tubing
- Unground neck to accept rubber stoppers
- Withstands repeated sterilization (wet or dry)
Specifications
2000 mL | |
Borosilicate Glass | |
Aspirator Bottle |
28 mm | |
248 mm | |
138 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction