Learn More
Pyrex™ Aspirator Bottle with Tubing Side Arm
Store and discharge a variety of liquids with this robust aspirator bottle. Pyrex™ Aspirator Bottles with Tubing Side Arm are useful as a delivery or storage container for a wide range of solutions. These bottles feature sturdy ring necks with a heavy duty rims.
Brand: Pyrex 1531/10D
258.40 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : NEW
Additional Details : Weight : 3.32000kg
Description
•Tubular side arm for attachment of 9mm bore flexible tubing•Unground neck to accept rubber stoppers•Can withstand repeated sterilization (wet or dry)Specifications
1000 mL | |
Borosilicate Glass | |
Aspirator Bottle |
28 mm | |
200 mm | |
111 mm |