missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Aspirator Bottle with Tubing Side Arm

Store and discharge a variety of liquids with this robust aspirator bottle. Pyrex™ Aspirator Bottles with Tubing Side Arm are useful as a delivery or storage container for a wide range of solutions. These bottles feature sturdy ring necks with a heavy duty rims.

Brand:  Pyrex 1531/08D

 View more versions of this product

Product Code. 11307884

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tubular side arm for attachment of 9mm bore flexible tubing
  • Unground neck to accept rubber stoppers
  • Withstands repeated sterilization (wet or dry)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mL
Borosilicate Glass
Aspirator Bottle
23 mm
163 mm
89 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers