missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Filter tube

Pyrex™ Borosilicate glass filter tubes are perfect for applications with Pyrex™ short stem funnels.

Brand:  Pyrex 2110/06M

Code : NEW

Additional Details : Weight : 0.39000kg

 View more versions of this product

Product Code. 15547690

  • £478.00 / Pack of 50
Estimated Shipment: 09-12-2024
to see stock.
Add to basket

Description

Description

Pyrex™ Borosilicate glass filter tubes are perfect for applications with Pyrex™ short stem funnels.

  • For small scale filtrations
  • Ideal for use with Pyrex™ short stem funnels
Specifications

Specifications

15mm
Pyrex Glass, Borosilicate Glass
Tube Filter
6mm
Short-stemmed funnel Pyrex™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers