missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Gooch Crucible with Integral Glass Sintered Disc

Ideal for drying precipitates to constant mass at 110°C. Pyrex™ Gooch Crucible includes an integral sintered disc.

Brand:  Pyrex 3660/02M

 View more versions of this product

Product Code. 10570101

  • £141.00 / Pack of 5
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for drying precipitates to constant mass at 110°C. PyrexTM Gooch Crucible includes an integral sintered disc.

  • Ideal for drying precipitates to constant mass at 110°C
  • Suitable for ignition work up to 450°C
  • With integral glass sintered disc
  • Manufactured from chemically resistant Pyrex™ borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications

Specifications

With Integral Glass sintered disc
Borosilicate Glass
450°C
53mm
1
30mm
Ideal for drying precipitates to constant mass at 110°C and for ignition work up to 450°C
30 mL
Gooch crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.