missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Gooch Crucible with Integral Glass Sintered Disc
Ideal for drying precipitates to constant mass at 110°C. Pyrex™ Gooch Crucible includes an integral sintered disc.
Brand: Pyrex 3650/02M
This item is not returnable.
View return policy
Description
Ideal for drying precipitates to constant mass at 110°C. PyrexTM Gooch Crucible includes an integral sintered disc.
- Ideal for drying precipitates to constant mass at 110°C
- Also suitable for ignition work up to 450°C (see safety notice)
- With integral glass sintered disc
- Manufactured from chemically resistant Pyrex™ borosilicate glass
Specifications
With Integral Glass sintered disc | |
Borosilicate Glass | |
450°C | |
45mm | |
1 |
20mm | |
Ideal for drying precipitates to constant mass at 110°C and for ignition work up to 450°C | |
10 mL | |
Gooch crucible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction