missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper
Store reagents, analytical standards and culture media in these glass reagent bottles. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper features excellent chemical resistance and can withstand high temperatures.
£39.50 - £2590.00
Specifications
Material | Borosilicate Glass |
---|---|
Certifications/Compliance | ISO 4796-2 |
Closure Included | Yes |
Mouth | Narrow |
Closure Material | Glass |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12643292
|
Pyrex
1521/04D |
100 mL |
£308.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
12653292
|
Pyrex
1521/06D |
250 mL |
£514.00
Pack of 10 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
11307864
|
Pyrex
1521/08D |
500 mL |
£439.00
Pack of 10 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
11317864
|
Pyrex
1521/10D |
1000 mL |
£555.00
Pack of 10 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
15537690
|
Pyrex
1521/12D |
2000 mL |
£2590.00
Pack of 30 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11337864
|
Pyrex
1521/16D |
5000 mL |
£183.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
11347864
|
Pyrex
1521/18D |
10,000 mL |
£292.00
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
11357864
|
Pyrex
1521/22D |
20,000 mL |
£606.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
11397854
|
Pyrex
1521/06D |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Complies with ISO 4796-2
- Supplied with solid glass stopper
- Pyrex™ borosilicate glass with ground neck
- Autoclavable
Specifications
Borosilicate Glass | |
Yes | |
Glass |
ISO 4796-2 | |
Narrow | |
Stopper |