Learn More
Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask
Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.
£51.40 - £251.00
Specifications
Certifications/Compliance | ISO 1773, DIN 12353 |
---|---|
Graduations | Yes |
Writing Surface | Yes |
Graduated | Yes |
Product Code | Brand | Diameter (Metric) Outer | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||
10348669
|
Pyrex
1130/12D |
79 mm | 200 mL | 131 mm |
£106.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||
10460160
|
Pyrex
1130/32D |
187 mm | 3000 mL | 310 mm |
£142.00
Pack of 2 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10765332
|
Pyrex
1130/36D |
220 mm | 5 L | 365 mm |
£177.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11972568
|
Pyrex
1130/11M |
42 mm | 25 mL | 70 mm |
£51.40
Pack of 10 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
11962568
|
Pyrex
1130/06D |
64 mm | 100 mL | 105 mm |
£123.00
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||
11961566
|
Pyrex
1130/30D |
166 mm | 2000 mL | 280 mm |
£251.00
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11982568
|
Pyrex
1130/02M |
51 mm | 50 mL | 85 mm |
£121.00
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12579105
|
Pyrex
1130/03 |
25 mm | 5 mL | 52 mm |
£114.00
Pack of 10 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||
12589105
|
Pyrex
1130/05M |
30 mm | 10 mL | 60 mm |
£53.15
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
11902619
|
Pyrex
1130/14D |
85 mm | 250 mL | 145 mm |
£147.58
Pack of 10 |
In Stock
Log in to see stock. |
|||
11912619
|
Pyrex
1130/20D |
105 mm | 500 mL | 180 mm |
£176.00
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||
11922619
|
Pyrex
1130/26D |
131 mm | 1000 mL | 220 mm |
£243.00
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||
11910905
|
Pyrex
1130/10D |
74 mm | 150 mL | 118 mm |
£135.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||
12519115
|
Pyrex
1130/16D |
87 mm | 300 mL | 156 mm |
£92.30
Pack of 10 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
15685767
|
Pyrex
1130/08D |
Body: 69 mm | 125 mL | 117 mm |
£97.65
Pack of 10 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
Description
Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.
- Complies with DIN ISO 1773
- Erlenmeyer pattern
- Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
- Graduated to full capacity
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
Specifications
ISO 1773, DIN 12353 | |
Yes | |
Yes | |
Yes |