Learn More
Pyrex™ Borosilicate Glass Plain Neck Kjeldahl Flask
Suitable for the determination of nitrogen
£197.00 - £446.00
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
11313624
|
Pyrex
1200/16M |
800 mL | 115 mm |
£446.00
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
11303624
|
Pyrex
1200/12D |
500 mL | 101 mm |
£245.00
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
11383614
|
Pyrex
1200/04M |
100 mL | Bulb: 58 mm |
£197.00
Pack of 10 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||||
15547680
|
Pyrex
1200/10D |
300 mL | 81 mm |
£445.00
Pack of 20 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
Description
- Withstands high temperatures
- Resistant to chemical corrosion
Specifications
Borosilicate Glass | |
Plain |