missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles

Product Code. 10230700
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.

Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.

  • Complies with ISO 4796-1
  • Manufactured from Pyrex™ borosilicate glass
  • Excellent resistance to chemical attack and high temperatures
  • Supplied with PBT screw caps that have a PTFE faced sealing disc
  • Autoclavable
  • White graduations and marking spot
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2000 mL
For Use With (Application) Ideal for storage of reagents, analytical standards and culture media etc
Material Borosilicate Glass
No. per Pack 10/PK
Closure Color Black
Certifications/Compliance ISO 4796-1
Closure Included Yes
Autoclavable Autoclavable
Graduated Yes
Height (Metric) 245 mm
Closure Liner PTFE
Closure Material Phenolic
Closure Size 60 mm
Diameter (Metric) 136 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'