missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Reagent Bottles
Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.
£114.00 - £2295.00
Specifications
For Use With (Application) | Ideal for storage of reagents, analytical standards and culture media etc |
---|---|
Material | Borosilicate Glass |
Closure Color | Black |
Certifications/Compliance | ISO 4796-1 |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11940326
|
Pyrex
1515/01D |
25 mL |
£162.00
Pack of 10 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
11961598
|
Pyrex
1515/02D |
50 mL | |||||||
10105044
|
Pyrex
1515/04D |
100 mL |
£167.18
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10411235
|
Pyrex
1515/06D |
250 mL |
£124.87
Pack of 10 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
10786885
|
Pyrex
1515/08D |
500 mL |
£228.07
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10422575
|
Pyrex
1515/10D |
1000 mL |
£246.65
Pack of 10 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||||
10230700
|
Pyrex
1515/12D |
2000 mL |
£795.00
Pack of 10 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||||
11347854
|
Pyrex
1515/16D |
5000 mL |
£180.00
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
11357854
|
Pyrex
1515/18D |
10,000 mL |
£302.00
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
15517690
|
Pyrex
1515/20D |
15,000 mL | |||||||
15527690
|
Pyrex
1515/22D |
20,000 mL |
£2295.00
Pack of 5 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
Description
Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.
- Complies with ISO 4796-1
- Manufactured from Pyrex™ borosilicate glass
- Excellent resistance to chemical attack and high temperatures
- Supplied with PBT screw caps that have a PTFE faced sealing disc
- Autoclavable
- White graduations and marking spot
Specifications
Ideal for storage of reagents, analytical standards and culture media etc | |
Black | |
Yes | |
Yes | |
Phenolic |
Borosilicate Glass | |
ISO 4796-1 | |
Autoclavable | |
PTFE | |
Screw Cap |