missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring

Product Code. 11982619
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20,000 mL
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10

Product Code. 11982619

Brand: Pyrex 1516/04D

Only null left
Add to basket
Edge
Add to basket

Mix and store a wide range of culture mediums with these glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140°C
TRUSTED_SUSTAINABILITY

Specifica

Capacity (Metric) 100 mL
Capacity (English) 3.4 oz.
Color Clear
Diameter (English) 2.2 in.
For Use With (Application) Ideal for mixing and storage of culture medium
Includes Polypropylene screw cap and pouring ring
Material Borosilicate Glass
Quantity 10 Pk.
Thread Size (Metric) GL 45
Certifications/Compliance ISO 4796
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Wide
Shape Round
Height (Metric) 100 mm
Product Type Reagent Bottle
For Use With (Equipment) Ideal for Mixing and Storage of Culture Medium
Height (English) 3.9 in.
Closure Material Polypropylene
Closure Size 45 mm
Diameter (Metric) 56 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato