missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Round Bottom Test Tubes

Versatile, round-bottom test tubes constructed of borosilicate. Pyrex™ Borosilicate Glass Round Bottom Test Tubes are supplied in packages of 100.

Brand:  Pyrex™ 612-20

Code : A0

Additional Details : Weight : 3.70000kg

Product Code. 12024617

  • £154.00 / Pack of 100
Estimated Shipment: 17-10-2024
to see stock.
Add to basket

Description

Description

  • Borosilicate glass construction
  • Supplied in packs of 100
Specifications

Specifications

40 mL
Borosilicate Glass
20 mm
200 mm
100 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers