missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Vacuum Desiccator
Designed for use under vacuum.
Brand: Pyrex 1594/02D
508.62 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed for use under vacuum
- Sturdy construction in borosilicate glass
- Ground glass flanges
- Cover has a 24/29 socket to accept desiccator stopcocks
- Choice of two sizes - to accommodate 150mm or 200mm diameter plates
- Plates are positioned on an internal ledge within the base
- Accessory metal perforated plate and Rotaflo™ glass/PTFE stopcock available
Specifications
Borosilicate Glass | |
24/29 | |
Flange: 170 mm |
Complete Dessicator | |
85 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction