Learn More
Pyrex™ Borosilicate Glass Watch Glasses
Product Code. 11383444
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
80 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Unit Size:
Each
Pack of 10
Product Code. 11383444
Brand: Pyrex 1060/09D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex™ borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Material Borosilicate Glass
Shape Round
Type Watch Glass
For Use With (Application) Evaporation Work and Covering Heated Beakers
Interior Finish Polished
Diameter (Metric) 60 mm