Learn More
Pyrex™ Wide Neck Round Bottom Boiling Flask
Made of borosilicate glass
£125.72 - £1030.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12539105
|
Pyrex
1120/08D |
250 mL |
£199.00
Pack of 10 |
Estimated Shipment: 17-03-2025
Log in to see stock. |
|||||
12549105
|
Pyrex
1120/14D |
500 mL |
£232.00
Pack of 10 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
12549975
|
Pyrex
1120/22D |
2000 mL |
£606.00
Pack of 10 |
Estimated Shipment: 17-03-2025
Log in to see stock. |
|||||
11991718
|
Pyrex
1120/29D |
6 L | N/A | ||||||
15567660
|
Pyrex
1120/04M |
100 mL |
£369.00
Pack of 30 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
15577660
|
Pyrex
1120/18D |
1000 mL |
£1030.00
Pack of 30 |
Estimated Shipment: 17-03-2025
Log in to see stock. |
|||||
15587660
|
Pyrex
1120/26D |
4 L | N/A | ||||||
Description
- Complies with ISO 24450 and DIN 12353
- With wide neck to facilitate addition of solid samples etc
- Resistant to chemical corrosion and thermal shock
Specifications
Borosilicate Glass | |
Round |