Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass with hexagonal glass.
£145.00 - £290.00
Specifications
Base Type Hexagonal
Type Graduated Cylinder
Material Glass
Certifications/Compliance ISO 4788
Class A
View More Specs
Products 8
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15707957
View Documents
Pyrex
3212/12WC
250 mL
£222.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15727947
View Documents
Pyrex
3212/04WC
10 mL
£146.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15727957
View Documents
Pyrex
3212/14WC
500 mL
£252.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15747947
View Documents
Pyrex
3212/06WC
25 mL
£198.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15747957
View Documents
Pyrex
3212/16WC
1000 mL
£290.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15767947
View Documents
Pyrex
3212/08WC
50 mL
£157.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15767957
View Documents
Pyrex
3212/18WC
2000 mL
£227.00
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15787947
View Documents
Pyrex
3212/10WC
100 mL
£145.00
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
Description
Description
  • Complies with ISO 4788
  • Calibrated to Class A tolerances
  • Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
  • Conformity H certified
  • Printed with individual serial number for batch certification
  • Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
  • Graduations in white enamel for improved visibility with dark colored solutions
  • Supplied with polyethylene stopper
Specifications
Specifications
Hexagonal
Glass
A
Polyethylene
Graduated Cylinder
ISO 4788
Polyethylene Stopper