missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Cloning Cylinders
Create identical cells by surrounding a single cell or group of cells
Brand: Corning™ 3166-8
This item is not returnable.
View return policy
Description
Clone a single cell or group of cells by surrounding them with this glass cylinder. Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an isolated well.

Specifications
Borosilicate Glass | |
8 mm | |
0.31 in. | |
0.31 in. |
8 mm | |
Non-sterile | |
Cloning Cylinder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ PYREX™ Cloning Cylinders > 8 mm
Spot an opportunity for improvement?Share a Content Correction