missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Conical Glass Funnel with PTFE Key

Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ Conical Glass Funnel with interchangeable PTFE stopcock. Complies with ISO 4800 Type 1 Standards.

Brand:  Pyrex 2190/01M

 View more versions of this product

Product Code. 12423461

  • £86.45 / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

    • Complies to ISO 4800 Type 1
    • Manufactured from chemically resistant Pyrex™ borosilicate glass
    • With interchangeable PTFE key
    • Supplied with both polyethylene and glass stoppers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL
ISO 4800
Glass stopper, ground Glass key
Separating funnel
Borosilicate Glass
Stopcock bore: 2mm
14/23
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers