missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Desiccator Stopcock
For use with Pyrex™ vacuum desiccators
Brand: Pyrex
Additional Details : Weight : 0.07000kg
Description
For use with Pyrex™ vacuum desiccators
- With Rotaflo™ stopcock for vacuum tight seal
- Contains a PTFE key and a 24/29 ground glass cone to fit the desiccator lids
Specifications
Desiccator Stopcock | |
Vacuum desiccators |