missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Funnel with Sintered Glass Disc

Product Code. p-8002586
Click to view available options
Capacity (Metric):
20 mL
30 mL
Porosity:
2
3
4
Diameter (Metric) Disc:
20mm
30mm
Height (Metric):
130 mm
144 mm
Unit Size:
Each
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 11313674

Brand: Pyrex 3730/06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper.

Manufactured from Pyrex™ Borosilicate Glass. Pyrex™ Funnel with Sintered Glass Disc is used for the filtration of solutions. Resistant to most corrosive agents, and eliminates the need for filter paper

  • Sintered disc manufactured from Pyrex™ borosilicate glass
  • For the filtration of solutions: - No requirement for filter paper - Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Available in a choice of porosities
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 20 mL
Diameter (Metric) Stem 8mm
Material Pyrex Glass, Borosilicate Glass
Height (Metric) 130 mm
For Use With (Application) For filtration of solutions
Diameter (Metric) Disc 20mm
Includes Sintered glass disc
Length (Metric) Stem 75mm
Porosity 3
Pore Size 16 to 40μm
Product Type Filter funnel
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.