Learn More
Pyrex™ Gas Distribution Tube
Click to view available options
Description:
Porosity: 0
Porosity: 1
Porosity: 2
Porosity: 3
Unit Size:
Pack of 2
Description
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
- Ideal for the dispersion of gas in liquids
- Borosilicate glass tube with sintered thimble
- Choice of thimble porosities to achieve optimum gas bubble size
Specifications
Specifications
| Description | Porosity: 3 |
| Material | Borosilicate Glass (3.3) |
| Overall Length (Metric) | 300 mm |
| For Use With (Application) | For the dispersion of gas in liquids |
| Includes | Sintered thimble |