Learn More
Pyrex™ Heavy Walled Test Tube
Pyrex™ Heavy Walled Test Tube is designed to provide extra mechanical strength.
Brand: Pyrex 1626/13M
135.75 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : NEW
Additional Details : Weight : 2.37000kg
Description
- Conforms to ISO 4142
- Rimless test tubes with a heavy wall to provide extra mechanical strength
- Robust construction reduces possibility of breakages: - Lower replacement costs - Improved user safety
- Manufactured from Pyrex™ borosilicate glass for high resistance to thermal shock and chemical attack
Specifications
Borosilicate Glass | |
100/Pk. | |
Test Tube |