missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Labware Polyethylene Reservoir Bottle
Feed liquids into a Sofnol burette with easy this filling squeeze bottle. The Pyrex™ labware polyethylene reservoir bottle is a replacement part for the Sofnol, auto zero, picnch clip, class B burette
£29.50 - £62.95
Specifications
For Use With (Equipment) | Sofnol Burette |
---|---|
Material | Polyethylene |
Type | Reservoir Bottle |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11343544
|
Pyrex
3299/06 |
1000 mL |
£62.95
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||||||
11333544
|
Pyrex
3299/04 |
500 mL |
£29.50
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||||||
Description
- Polyethylene
- Squeeze bottle
- For use with Sofnol burette
- Squeeze the bottle to fill the burette
- Release pressure to allow excess liquid to drain back into bottle and set the burrette to zero
- Used for liquid testing
Specifications
Sofnol Burette | |
Polyethylene | |
Reservoir Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Labware Polyethylene Reservoir Bottle