missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Media Bottle High-Temperature Caps, Open-Top

Product Code. 17233474
Click to view available options
Thread Size (Metric):
GL 25
GL 32
GL 45
This item is not returnable. View return policy
This item is not returnable. View return policy

With drip-free pouring ring

These are various sizes and types of screw caps for use with storage bottles.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material PBT
No. per Case 10/Cs.
No. per Pack 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 25
Closure Color Red
Product Type Open Top Cap
For Use With (Equipment) Storage bottle
Closure Material PBT
Closure Type High Temperature
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'