missing translation for 'onlineSavingsMsg'
Learn More
Pyrex™ Narrow-Mouth Reagent Bottle Product Code.: 11367874
Estimated Shipment: 25-03-2025
to see stock.
Add to basket

Pyrex™ Narrow-Mouth Reagent Bottle

Product Code. 11367874
10,000 mL
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
This item is not returnable. View return policy

Product Code. 11367874

Brand: Pyrex 1526/18D

Estimated Shipment: 25-03-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from Pyrex™ superior strength borosilicate glass. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles are ideal for storing chemicals in the laboratory. Use rubber stoppers (not included) to secure.

  • Unground neck for use with rubber stoppers (not supplied)
  • Borosilicate glass provides strong chemical resistance
  • Complies with ISO 4796
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10,000 mL
Material Borosilicate Glass
Neck ID (Metric) 59mm
Quantity Each
Neck Finish Unground
Certifications/Compliance ISO 4796
Closure Included No
Mouth Narrow
Height (Metric) 398 mm
Diameter (Metric) 227 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex™ Narrow-Mouth Reagent Bottle >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.