missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Narrow-Mouth Reagent Bottle
550.24 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
Description
Manufactured from Pyrex™ superior strength borosilicate glass. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles are ideal for storing chemicals in the laboratory. Use rubber stoppers (not included) to secure.
- Unground neck for use with rubber stoppers (not supplied)
- Borosilicate glass provides strong chemical resistance
- Complies with ISO 4796
Specifications
Capacity (Metric) | 20,000 mL |
Material | Borosilicate Glass |
Neck ID (Metric) | 59mm |
Quantity | Each |
Neck Finish | Unground |
Certifications/Compliance | ISO 4796 |
Closure Included | No |
Mouth | Narrow |
Height (Metric) | 492 mm |
Diameter (Metric) | 288 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'