Learn More
Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap
Manufactured from Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage. Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap reduces the hazard of flying glass and liquid splashes in the event of breakage. Coating withstands steam sterilisation up to 135° C. Bottles are supplied without cap or pouring ring.
£166.00 - £969.00
Specifications
Coating | Plastic |
---|---|
Color | Clear |
For Use With (Application) | Ideal for mixing and storage of culture medium |
Material | Borosilicate Glass |
Certifications/Compliance | ISO 4796 |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
12539135
|
Pyrex
1518/04 |
100 mL |
£166.00
Pack of 10 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11962629
|
Pyrex
1518/06 |
250 mL | N/A | |||||||
12069012
|
Pyrex
1518/08 |
500 mL | N/A | |||||||
12559135
|
Pyrex
1518/10 |
1000 mL | N/A | |||||||
10221560
|
Pyrex
1518/12 |
2000 mL | N/A | |||||||
10596340
|
Pyrex
1518/16 |
5000 mL | N/A | |||||||
10308859
|
Pyrex
1518/18 |
10,000 mL |
£969.00
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
Description
Manufactured from Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage. Pyrex™ Plastic Coated Borosilicate Glass Wide Neck Round Media Bottles Without Cap reduces the hazard of flying glass and liquid splashes in the event of breakage. Coating withstands steam sterilisation up to 135° C. Bottles are supplied without cap or pouring ring.
- Bottles supplied without cap or pouring ring
- Borosilicate glass with clear plastic coating to provide added protection against mechanical breakage
- Reduces the hazard of flying glass and liquid splashes in the event of breakage
- Printed with retrace code: downloadable batch certificate
- Coating withstands steam sterilisation up to 135°C
Specifications
Plastic | |
Ideal for mixing and storage of culture medium | |
ISO 4796 | |
Yes | |
Round |
Clear | |
Borosilicate Glass | |
Autoclavable | |
Wide | |
45 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.