missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Scoop with Hollow Stem

Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.

Brand:  Pyrex 2115/04M

230.41 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11970885

  • £367.00 / Pack of 10
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.

  • With hollow stem to aid introduction of the powder into narrow neck vessels e.g., volumetric flasks
  • The flat base ensures stability on the balance pan
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 mL
Borosilicate Glass
85 mm
Weighing scoop
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.