missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key

Supplied with hollow glass stopper and interchangeable PTFE key for grease-free performance. Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key provides good control of the liquid/liquid interface.

Brand:  Pyrex 443060N

 View more versions of this product

Product Code. 12579165

  • £180.00 / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gilson style with long delivery stem for good control of the liquid/liquid interface
  • With interchangeable PTFE key for grease-free performance
  • Supplied with hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1000 mL
Hollow Glass stopper, PTFE key
(Bore Size) 4mm
Borosilicate Glass
29/32
Separatory Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers