Learn More
Pyrex™ Short Stem Glass Funnel
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
£17.75 - £302.00
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Diameter (Metric) Disc | Length (Metric) Stem | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Diameter (Metric) Disc | Length (Metric) Stem | Price | Quantity & Availability | ||
12529945
|
Pyrex
2140/03M |
70mm | 125 mm | 70 to 90mm (Filter paper size) | 75mm |
£273.00
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||
12595067
|
Pyrex
2140/06M |
80mm | 144 mm | 90 to 150mm (Filter paper size) | 82mm |
£299.28
Pack of 10 |
In Stock
Log in to see stock. |
||
12099907
|
Pyrex
2140/11M |
125mm | 220 mm | 185 to 240mm (Filter paper size) | 122mm |
£62.80
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
11307944
|
Pyrex
2140/14M |
200mm | 345 mm | 320 to 385mm (Filter paper size) | 193mm |
£78.10
Each |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||
11397934
|
Pyrex
2140/12M |
150mm | 270 mm | 240 to 270mm (Filter paper size) | 154mm |
£90.35
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
12585067
|
Pyrex
2140/01M |
35mm | 60 mm | 45 to 55mm (Filter paper size) | 35mm |
£230.00
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||
11347934
|
Pyrex
2140/02M |
45mm | 78 mm | 55 to 70mm (Filter paper size) | 45mm |
£238.39
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||
11992568
|
Pyrex
2140/10M |
105mm | 186 mm | 150 to 185mm (Filter paper size) | 106mm |
£302.00
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||
11357934
|
Pyrex
2140/03M |
70mm | 125 mm | 70 to 90mm (Filter paper size) | 75mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12923611
|
Pyrex
2140/02M |
45mm | 78 mm | 55 to 70mm (Filter paper size) | 45mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11367934
|
Pyrex
2140/06M |
80mm | 144 mm | 90 to 150mm (Filter paper size) | 82mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11337934
|
Pyrex
2140/01M |
35mm | 60 mm | 45 to 55mm (Filter paper size) | 35mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
- With 60° bowl angle for precise fitting of filter paper
- Manufactured from chemical and heat resistant Pyrex™ borosilicate glass
Spécification
NF B 35 032 | |
Short stem funnel |