missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Spare Stopcock Key

Pyrex™ Spare Stopcock Key is a fully interchangeable replacement PTFE stopcock key, supplied complete with retaining device.

Brand:  Pyrex SST105K

117.15 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15718067

  • £123.00 / Pack of 5
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fully interchangeable PTFE keys for Pyrex™ stopcocks
  • Supplied complete with plastic screw thread retaining device
  • Keys are self lubricating, avoiding the need for stopcock grease
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

18.8 mm stopcock
4.0 mm bore
5/Pk
PTFE
Straight Bore
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex™ Spare Stopcock Key > Diameter: 18.8 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.