missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ VISTA™ Petri Culture Dishes

These flat, clear Pyrex Vista dishes will withstand repeated sterilization (wet or dry). The edges are beaded to provide greater mechanical strength.

Brand:  Corning™ 70165-60

Additional Details : Weight : 1.58170kg

 View more versions of this product

Product Code. 17273564

  • £132.86 / Pack of 24
Estimated Shipment: 11-12-2024
to see stock.
Add to basket

Description

Description

These flat, clear dishes are made from soda lime silica glass and will withstand repeated sterilization (wet or dry). The edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, thereby reducing the capillary action of the condensed moisture on the sides. They are not affected chemically or thermally by any of the methods commonly employed in laboratories where sterilization is routinely used. The covered dish is not airtight. Bottoms also have a triangular enamel reference point for serial dilutions.

Specifications

Specifications

Clear
Round
Beaded
12/Pk.
60 mm
Soda Lime Glass
Cell Culture Petri Dish
24/Cs.
15 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers