missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-2-boronic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Pyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.
Notes
Incompatible with strong oxidizing agents.
Pyridine-2-boronic acid is used as a key starting material for the formation of pyridyl boronic ester such as pyridine-2-boronic acid dimethyl ester. Furthermore, the dimethyl ester reacts with bromquinoline to prepare the 2-pyridyl derivative of quinoline.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Quantity | 250 mg |
Formula Weight | 122.92 |
Percent Purity | 95% |
Chemical Name or Material | Pyridine-2-boronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'