missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-boronic acid 1,3-propanediol ester, 98+%, Thermo Scientific Chemicals

Product Code. 11303906
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131534-65-1
Molecular Formula C8H10BNO2
Molecular Weight (g/mol) 162.98
MDL Number MFCD01075749
InChI Key ZUUZNOFTKGDLLN-UHFFFAOYSA-N
Synonym 3-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid 1,3-propanediol ester, pyridine-3-boronic acid 1,3-propanediol cyclic ester, 1,3-propanediol cyclic ester, 2-3-pyridyl-1,3,2-dioxaborinane, 3-1,3,2 dioxaborinan-2-yl-pyridine, pyridine-3-boronic acid 1,3-propanediol ester, 3-pyridineboronic acid glycol ester, 2-3-pyridyl-1,3,2-dioxaborane, 2-pyridin-3-yl-1,3,2-dioxaborinane
PubChem CID 4198005
IUPAC Name 3-(1,3,2-dioxaborinan-2-yl)pyridine
SMILES C1COB(OC1)C1=CC=CN=C1

Specifications

Melting Point 100°C to 104°C
Quantity 5 g
Beilstein 6138748
Formula Weight 162.98
Percent Purity ≥98%
Chemical Name or Material Pyridine-3-boronic acid 1,3-propanediol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'