missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid, 97%, Thermo Scientific™

£23.00 - £187.00

Chemical Identifiers

CAS 1692-15-5
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD01074545
InChI Key QLULGIRFKAWHOJ-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
PubChem CID 2734379
SMILES OB(O)C1=CC=NC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10337853
View Documents
Thermo Scientific Maybridge
CC04212DA
1 g Amber Glass Bottle
£23.00
1g
Please to purchase this item. Need a web account? Register with us today!
10503153
View Documents
Thermo Scientific Maybridge
CC04212EA
10 g Amber Glass Bottle
£187.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1692-15-5
122.92
QLULGIRFKAWHOJ-UHFFFAOYSA-N
2734379
C5H6BNO2
MFCD01074545
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
OB(O)C1=CC=NC=C1

Specifications

1692-15-5
C5H6BNO2
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
OB(O)C1=CC=NC=C1
122.92
122.92
Pyridin-4-ylboronic acid
97%
MFCD01074545
QLULGIRFKAWHOJ-UHFFFAOYSA-N
(pyridin-4-yl)boronic acid
2734379
97%
Videos
SDS
Documents

Documents

Product Certifications