missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-4-boronic acid hydrochloride, 95%, Thermo Scientific Chemicals
£33.50 - £124.00
Chemical Identifiers
CAS | 913835-65-1 |
---|---|
Molecular Formula | C5H7BClNO2 |
Molecular Weight (g/mol) | 159.38 |
MDL Number | MFCD06201888 |
InChI Key | NCZXYQJUECNKAD-UHFFFAOYSA-N |
Synonym | pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl |
PubChem CID | 44118740 |
IUPAC Name | pyridin-4-ylboronic acid;hydrochloride |
SMILES | Cl.OB(O)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479397
|
Thermo Scientific Alfa Aesar
H52648.03 |
1 g |
£33.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15489397
|
Thermo Scientific Alfa Aesar
H52648.06 |
5 g |
£124.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
913835-65-1 | |
159.38 | |
NCZXYQJUECNKAD-UHFFFAOYSA-N | |
44118740 | |
Cl.OB(O)C1=CC=NC=C1 |
C5H7BClNO2 | |
MFCD06201888 | |
pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl | |
pyridin-4-ylboronic acid;hydrochloride |
Specifications
913835-65-1 | |
C5H7BClNO2 | |
pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl | |
Cl.OB(O)C1=CC=NC=C1 | |
159.38 | |
159.38 | |
Pyridine-4-boronic acid hydrochloride |
340°C | |
MFCD06201888 | |
NCZXYQJUECNKAD-UHFFFAOYSA-N | |
pyridin-4-ylboronic acid;hydrochloride | |
44118740 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only