Learn More
Pyridine, 99+%, Thermo Scientific Chemicals
£32.68 - £345.72
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464054
|
Thermo Scientific Alfa Aesar
A12005.AP |
500 mL |
£32.68
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474054
|
Thermo Scientific Alfa Aesar
A12005.0F |
2500 mL |
£110.55
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484054
|
Thermo Scientific Alfa Aesar
A12005.0D |
10,000 mL |
£345.72
10000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane and pyridine-sulfur trioxide are widely used owing to their superior characteristics in their reactivity and solubility characteristics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAs polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane and pyridine-sulfur trioxide are widely used owing to their superior characteristics in their reactivity and solubility characteristics.
Solubility
Miscible with water, alcohol, ether, pet ether, and oils.
Notes
Pyridine is stable under normal temperatures and pressures. It is incompatible with strong oxidizing agents, and acids.
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
°C to 42°C | |
0.978 | |
20°C (68°F) | |
1.51 | |
C5H5N | |
UN1282 | |
14,7970 | |
Miscible with water,alcohol,ether,pet ether,and oils. | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
≥99% |
110-86-1 | |
115°C | |
Liquid | |
≥99% | |
MFCD00011732 | |
103233 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1 | |
Pyridine |
Safety and Handling
GHS H Statement
H225-H302-H312-H332
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H312+H332-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
missing translation for 'einecsNumber' : 203-809-9
missing translation for 'rtecsNumber' : UR8400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Pyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : pyridine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.