missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, ≥99%, Reagent Grade, Honeywell™

Product Code. 15638040 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 15638040

Brand: Honeywell Research Chemicals P575061L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

pH 8.8 (at 20°C)
Melting Point -42°C
Color Colorless
Density 0.978g/mL at 25°C
Boiling Point 115°C (1013 hPa)
Quantity 1 L
Explosion Limit 0.124
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Odor Unpleasant
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 10mmHg (13.2°C), 20mmHg (25°C)
UN Number UN1282
Beilstein 103233
Solubility Information Fully miscible in water
Formula Weight 79.1g/mol
Percent Purity ≥99%
Chemical Name or Material Pyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.