missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, ≥99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
CAS | 110-86-1 |
Molecular Formula | C5H5N |
MDL Number | MFCD00011732 |
UN Number | UN1282 |
Beilstein | 103233 |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
SMILES | C1=CC=NC=C1 |
IUPAC Name | pyridine |
Molecular Weight (g/mol) | 79.102 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'