Learn More
Pyridine-d{5}, 99.5% (Isotopic), Thermo Scientific Chemicals
£30.15 - £114.57
Chemical Identifiers
CAS | 7291-22-7 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 84.13 |
MDL Number | MFCD00044639 |
InChI Key | JUJWROOIHBZHMG-RALIUCGRSA-N |
Synonym | pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c |
PubChem CID | 558519 |
IUPAC Name | 2,3,4,5,6-pentadeuteriopyridine |
SMILES | [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387928
|
Thermo Scientific Alfa Aesar
036471.04 |
2 g |
£30.15
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397928
|
Thermo Scientific Alfa Aesar
036471.09 |
10 g |
£114.57
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyridine-d{5} is used in the synthesis complexes with cobalt(II) halides, pyridine-d5, a deuterated form of pyridine, ionic liquid matrices (ILMs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7291-22-7 | |
84.13 | |
JUJWROOIHBZHMG-RALIUCGRSA-N | |
558519 | |
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H] |
C5H5N | |
MFCD00044639 | |
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c | |
2,3,4,5,6-pentadeuteriopyridine |
Specifications
Liquid | |
20°C (68°F) | |
1.506 | |
-42°C | |
C5H5N | |
UN1282 | |
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c | |
JUJWROOIHBZHMG-RALIUCGRSA-N | |
2,3,4,5,6-pentadeuteriopyridine | |
558519 | |
99.5% |
1.05 g/mL | |
Unpleasant | |
7291-22-7 | |
115°C | |
MFCD00044639 | |
114377 | |
Soluble in water,alcohol,ether,acetone,and benzene. | |
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H] | |
84.13 | |
84.14 | |
Pyridine-d5, Isotopic |
Safety and Handling
GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H312+H332-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
missing translation for 'einecsNumber' : 203-720-2
missing translation for 'tsca' : No
Recommended Storage : Ambient
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.