missing translation for 'onlineSavingsMsg'
Learn More
Pyridine, for HPLC Product Code.: 10791895

Pyridine, for HPLC

Product Code. 10791895
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10791895

Brand: Thermo Scientific Acros 396800010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-86-1 | C5H5N | 79.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -42.0°C
Absorbance 0.02 max. (1 cm cell vs HPLC-grade water) at 345nm, 0.01 max. (1 cm cell vs HPLC-grade water) at 400nm, 0.05 max. (1cm cell vs HPLC-grade water) at 340nm
CAS 110-86-1
Density 0.9780g/mL
Boiling Point 115.0°C to 116.0°C
Quantity 1 L
Flash Point 17°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.5085 to 1.5105
Residue after Evaporation 0.001% max.
Specific Gravity 0.978
Molecular Formula C5H5N
MDL Number MFCD00011732
UN Number 1282
Viscosity 0.95 mPa.s (20°C)
Water 0.05% max.
Merck Index 15, 8081
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1
Percent Purity ≥99.8%
Grade HPLC
Chemical Name or Material Pyridine, For HPLC
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463558 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog Number396800010
Lot NumberA0458200 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number396800010
Lot NumberA0453031 Certificate TypeCertificate of Analysis Date10/05/2023 Catalog Number396800010
Lot NumberA0447010 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number396800010
Lot NumberA0442996 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog Number396800010
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine, for HPLC >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.