missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide perbromide, tech. 90%, Thermo Scientific Chemicals

£33.50 - £219.00

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H5N·HBr3
MDL Number MFCD00013223
Synonym Pyridinium bromide perbromide; Pyridinium tribromide
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11499160
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15684.14
25 g
£33.50
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11409170
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15684.22
100 g
£57.40
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11419170
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15684.36
500 g
£219.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pyridine hydrobromide perbromide is used as a brominating reagent in alfa-bromination and alfa-thiocyanation of ketones, phenols, unsaturated and aromatic ethers. It is used as a raw material in the preparation of beta-adrenergic blocking agents. Furthermore, it is used as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide perbromide is used as a brominating reagent in alfa-bromination and alfa-thiocyanation of ketones, phenols, unsaturated and aromatic ethers. It is used as a raw material in the preparation of beta-adrenergic blocking agents. Furthermore, it is used as an analytical reagent.

Solubility
Soluble in methanol, acetic acid, ethanol, n-butanol and terahydrofuran. Insoluble in water, carbon tetrachloride, ethyl bromide, benzene, toluene, ligroin, petroleum ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

39416-48-3
C5H5N·HBr3
MFCD00013223
Pyridinium bromide perbromide; Pyridinium tribromide

Specifications

39416-48-3
C5H5N·HBr3
UN3261
14,7973
Soluble in methanol,acetic acid,ethanol,n-butanol and terahydrofuran. Insoluble in water,carbon tetrachloride,ethyl bromide,benzene,toluene,ligroin,petroleum ether.
90%
Pyridine hydrobromide perbromide
128°C to 136°C
MFCD00013223
3690144
Pyridinium bromide perbromide; Pyridinium tribromide
319.82
Technical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 254-446-8

missing translation for 'tsca' : Yes

Product Identifier
  • Pyridine hydrobromide perbromide

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only