missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11404223
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

Solubility
Soluble in water (85 g/100 ml) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-13-7
Molecular Formula C5H6ClN
Molecular Weight (g/mol) 115.56
MDL Number MFCD00012802
InChI Key AOJFQRQNPXYVLM-UHFFFAOYSA-N
Synonym pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
PubChem CID 10176127
IUPAC Name pyridine;hydrochloride
SMILES C1=CC=NC=C1.Cl

Specifications

Melting Point ∼145°C
Boiling Point 222°C to 224°C
Quantity 50 g
Beilstein 3615340
Sensitivity Hygroscopic
Solubility Information Soluble in water (85g/100 ml) and methanol.
Formula Weight 115.56
Percent Purity 98%
Chemical Name or Material Pyridine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'