missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, puriss. p.a., ACS reagent, ≥99.5%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
200 kg
Packaging:
Metal Drum
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
CAS | 110-86-1 |
CAS Max % | 100.0000% |
Molecular Formula | C5H5N |
Linear Formula | C21H14Br4O5S |
MDL Number | MFCD00005874 |
UN Number | UN1282 |
Beilstein | 103233 |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
SMILES | C1=CC=NC=C1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'