Learn More
Pyridoxal hydrochloride, 99%, Thermo Scientific Chemicals
£43.10 - £156.00
Chemical Identifiers
CAS | 65-22-5 |
---|---|
Molecular Formula | C8H10ClNO3 |
Molecular Weight (g/mol) | 203.622 |
MDL Number | MFCD00012809 |
InChI Key | FCHXJFJNDJXENQ-UHFFFAOYSA-N |
Synonym | pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride |
PubChem CID | 6171 |
IUPAC Name | 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride |
SMILES | CC1=NC=C(C(=C1O)C=O)CO.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423458
|
Thermo Scientific Alfa Aesar
A17855.06 |
5 g |
£43.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433458
|
Thermo Scientific Alfa Aesar
A17855.14 |
25 g |
£156.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.
Solubility
Soluble in water and ethanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids, strong reducing agents and strong bases. Store in a cool place.
Chemical Identifiers
65-22-5 | |
203.622 | |
FCHXJFJNDJXENQ-UHFFFAOYSA-N | |
6171 | |
CC1=NC=C(C(=C1O)C=O)CO.Cl |
C8H10ClNO3 | |
MFCD00012809 | |
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride | |
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride |
Specifications
65-22-5 | |
Odorless | |
MFCD00012809 | |
Hygroscopic | |
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride | |
FCHXJFJNDJXENQ-UHFFFAOYSA-N | |
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride | |
6171 | |
99% |
172°C to 174°C (decomposition) | |
C8H10ClNO3 | |
3656994 | |
14,7978 | |
Soluble in water and ethanol. | |
CC1=NC=C(C(=C1O)C=O)CO.Cl | |
203.622 | |
203.63 | |
Pyridoxal hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 200-602-5
missing translation for 'rtecsNumber' : UV1225000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only