missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine hydrochloride, 99%

Product Code. 11469912
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11469912

Brand: Thermo Scientific Alfa Aesar A12041.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58-56-0 | C8H12ClNO3 | 205.638 g/mol

Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.

Solubility
Soluble in water and alcohol. Slightly soluble in acetone. In soluble in ether and chloroform.

Notes
Light sensitive. Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
MDL Number MFCD00012807
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl

Specifications

Melting Point ∼206°C (decomposition)
pH 3
Odor Odorless
Quantity 500 g
Beilstein 3632435
Sensitivity Light sensitive
Merck Index 14,7982
Solubility Information Soluble in water and alcohol. Slightly soluble in acetone. In soluble in ether and chloroform.
Formula Weight 205.64
Percent Purity 99%
Chemical Name or Material Pyridoxine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.