missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15578114
Click to view available options
Quantity:
5 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 100 g
Formula Weight 205.64
Grade FCC/USP
Chemical Name or Material Pyridoxine Hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'