missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrogallol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 131.0°C to 135.0°C |
Color | White |
Boiling Point | 309.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Plastic bottle |
Linear Formula | C6H3(OH)3 |
Quantity | 100 g |
Beilstein | 06,1071 |
Merck Index | 15,8112 |
Show More |
Safety and Handling
Product Identifier
- Pyrogallol
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'